Merck
  • Possible contribution of epigenetic changes in the development of schizophrenia-like behavior in vasopressin-deficient Brattleboro rats.
Possible contribution of epigenetic changes in the development of schizophrenia-like behavior in vasopressin-deficient Brattleboro rats.
Behavioural brain research (2015-12-26)
Kornél Demeter, Bibiána Török, Anna Fodor, János Varga, Szilamér Ferenczi, Krisztina J Kovács, Ildikó Eszik, Viktor Szegedi, Dóra Zelena
ABSTRACT
Schizophrenia-like symptoms were detected in vasopressin-deficient (di/di) Brattleboro rats, and it was also suggested that schizophrenia might have an epigenetic component. We aimed to clarify if epigenetic changes contribute to schizophrenia-like behavior of this strain. Behavioral (locomotion by telemetry, cognition by novel object recognition, social recognition and social avoidance test, attention by pre-pulse inhibition) and epigenetic differences were compared between wild type and di/di animals. DNA methyltransferase1 (DNMT1), DNMT3a, as well as COMT, GAD, VGLUT1, 5HT2A, BDNF mRNA levels in prefrontal brain region and hippocampus were studied by qRT-PCR. Histone3 (H3) and H4 acetylation (Ac) were studied by western-blot followed by region specific examination of H3 lysine9 (K9) acetylation by immunohistochemistry. Impaired cognitive, social and attention behavior of di/di rats confirmed schizophrenia-like symptoms in our local colony. The pan-AcH3 immunoreactivity was lower in prefrontal region and elevated in the hippocampus of di/di animals. We found lower immunopositive cell number in the dorsal peduncular prefrontal cortex and the ventral lateral septum and increased AcH3K9 immunoreactivity in CA1 region of di/di animals. There were no major significant alterations in the studied mRNA levels. We confirmed that Brattleboro rat is a good preclinical model of schizophrenia. Its schizophrenia-like behavioral alteration was accompanied by changes in H3 acetylation in the prefrontal region and hippocampus. This may contribute to disturbances of many schizophrenia-related substances leading to development of schizophrenia-like symptoms. Our studies confirmed that not a single gene, rather fine changes in an array of molecules are responsible for the majority of schizophrenia cases.
Sigma-Aldrich
Anti-acetyl-Histone H4 (Lys12) Antibody, serum, Upstate®
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit