Merck
Copeptin as a serum biomarker of febrile seizures.
PloS one (2015-04-22)
Benjamin Stöcklin, Sotirios Fouzas, Paula Schillinger, Sevgi Cayir, Roswitha Skendaj, Michel Ramser, Peter Weber, Sven Wellmann
ABSTRACT
Accurate diagnosis of febrile seizures in children presenting after paroxysmal episodes associated with fever, is hampered by the lack of objective postictal biomarkers. The aim of our study was to investigate whether FS are associated with increased levels of serum copeptin, a robust marker of arginine vasopressin secretion. This was a prospective emergency-setting cross-sectional study of 161 children between six months and five years of age. Of these, 83 were diagnosed with febrile seizures, 69 had a febrile infection without seizures and nine had epileptic seizures not triggered by infection. Serum copeptin and prolactin levels were measured in addition to standard clinical, neurophysiological, and laboratory assessment. NCT01884766. Circulating copeptin was significantly higher in children with febrile seizures (median [interquartile range] 18.9 pmol/L [8.5-36.6]) compared to febrile controls (5.6 pmol/L [4.1-9.4]; p < 0.001), with no differences between febrile and epileptic seizures (21.4 pmol/L [16.1-46.6]; p = 0.728). In a multivariable regression model, seizures were the major determinant of serum copeptin (beta 0.509; p < 0.001), independently of clinical and baseline laboratory indices. The area under the receiver operating curve for copeptin was 0.824 (95% CI 0.753-0.881), significantly higher compared to prolactin (0.667 [0.585-0.742]; p < 0.001). The diagnostic accuracy of copeptin increased with decreasing time elapsed since the convulsive event (at 120 min: 0.879 [0.806-0.932] and at <60 min: 0.975 [0.913-0.997]). Circulating copeptin has high diagnostic accuracy in febrile seizures and may be a useful adjunct for accurately diagnosing postictal states in the emergency setting.