Merck
  • Acid phosphatase activity may affect the tuber swelling by partially regulating sucrose-mediated sugar resorption in potato.
Acid phosphatase activity may affect the tuber swelling by partially regulating sucrose-mediated sugar resorption in potato.
Journal of integrative plant biology (2008-08-21)
Da-Yong Wang, Yong Lian, De-Wei Zhu
ABSTRACT
APase activity is involved in regulating many physiological and developmental events by affecting the resorption process. In this study, we investigate the role of APase activity in tuber development in potato. APase activities were mainly localized in cytoplasm, gaps among cells and stroma of amyloplasts of parenchyma cells at the stage of tuber swelling. AP1, encoding a putative APase, was also highly expressed in swelling tubers and a low level of expression was observed in elongated stolons and matured tubers. Inhibition of APase activity by applying Brefeldin A, an inhibitor of APase production and secretion, significantly suppressed the tuber swelling and moderately affected the stolon elongation and the tuberization frequency. During tuber development, sucrose serves as the main soluble sugar for long-distance transportation and resorption. Moreover, inhibition of APase activity by Brefeldin A markedly reduced the sucrose content in tubers and further decreased the starch accumulation, suggesting that the function of APase in regulating the tuber swelling might be at least partially mediated by the sugar resorption. Exogenous sucrose treatments further indicate the important role of sucrose-mediated sugar resorption in tuber swelling. These results suggest that the APase activity might affect the tuber swelling by partially regulating the sucrose-mediated sugar resorption.
Product Description
Roche
Acid Phosphatase, grade II, from potato