Merck
  • A Study of the Wound Healing Mechanism of a Traditional Chinese Medicine, Angelica sinensis, Using a Proteomic Approach.
A Study of the Wound Healing Mechanism of a Traditional Chinese Medicine, Angelica sinensis, Using a Proteomic Approach.
Evidence-based complementary and alternative medicine : eCAM (2012-04-27)
Chia-Yen Hsiao, Ching-Yi Hung, Tung-Hu Tsai, Kin-Fu Chak
ABSTRACT
Angelica sinensis (AS) is a traditional Chinese herbal medicine that has been formulated clinically to treat various form of skin trauma and to help wound healing. However, the mechanism by which it works remains a mystery. In this study we have established a new platform to evaluate the pharmacological effects of total AS herbal extracts as well as its major active component, ferulic acid (FA), using proteomic and biochemical analysis. Cytotoxic and proliferation-promoting concentrations of AS ethanol extracts (AS extract) and FA were tested, and then the cell extracts were subject to 2D PAGE analysis. We found 51 differentially expressed protein spots, and these were identified by mass spectrometry. Furthermore, biomolecular assays, involving collagen secretion, migration, and ROS measurements, gave results that are consistent with the proteomic analysis. In this work, we have demonstrated a whole range of pharmacological effects associated with Angelica sinensis that might be beneficial when developing a wound healing pharmaceutical formulation for the herbal medicine.
Sigma-Aldrich
Anti-STMN1 Rabbit pAb, liquid, Calbiochem®