Merck
  • Farnesol-induced cell death and stimulation of 3-hydroxy-3-methylglutaryl-coenzyme A reductase activity in tobacco cv bright yellow-2 cells.
Farnesol-induced cell death and stimulation of 3-hydroxy-3-methylglutaryl-coenzyme A reductase activity in tobacco cv bright yellow-2 cells.
Plant physiology (2000-08-12)
A Hemmerlin, T J Bach
ABSTRACT
Growth inhibition of tobacco (Nicotiana tabacum L. cv Bright Yellow-2) cells by mevinolin, a specific inhibitor of 3-hydroxy-3-methylglutaryl-coenzyme A reductase (HMGR) could be partially overcome by the addition of farnesol. However, farnesol alone inhibited cell division and growth as measured by determination of fresh weight increase. When 7-d-old tobacco cv Bright Yellow-2 cells were diluted 40-fold into fresh culture, the cells exhibited a dose-dependent sensitivity to farnesol, with 25 microM sufficient to cause 100% cell death, as measured by different staining techniques, cytometry, and monitoring of fragmentation of genomic DNA. Cells were less sensitive to the effects of farnesol when diluted only 4-fold. Farnesol was absorbed by the cells, as examined by [1-(3)H]farnesol uptake, with a greater relative enrichment by the more diluted cells. Both mevinolin and farnesol treatments stimulated apparent HMGR activity. The stimulation by farnesol was also reflected in corresponding changes in the steady-state levels of HMGR mRNA and enzyme protein with respect to HMGR gene expression and enzyme protein accumulation.