Merck
  • Super-resolution microscopy reveals γ-secretase at both sides of the neuronal synapse.
Super-resolution microscopy reveals γ-secretase at both sides of the neuronal synapse.
Acta neuropathologica communications (2016-04-03)
Sophia Schedin-Weiss, Ina Caesar, Bengt Winblad, Hans Blom, Lars O Tjernberg
ABSTRACT
The transmembrane protein assembly γ-secretase is a key protease in regulated intramembrane processing (RIP) of around 100 type-1 transmembrane proteins. Importantly, it has a pathological role in Alzheimer disease (AD) as it generates the neurotoxic amyloid β-peptide from the amyloid precursor protein (APP). Studies on γ-secretase location are therefore crucial both from a biological and a therapeutic perspective. Despite several years of efforts in many laboratories, it is not clear where in the neuron γ-secretase exerts it's activities. Technical challenges include the fact that the active enzyme contains four protein components and that most subcellular compartments cannot be spatially resolved by traditional light microscopy. Here, we have used a powerful combination of the two nanoscopy techniques STORM and STED microscopy to visualize the location of γ-secretase in neurons using an active-site specific probe, with a focus on the synapse. We show that γ-secretase is present in both the pre-and postsynaptic compartments. We further show that the enzyme is enriched very close to the synaptic cleft in the postsynaptic membrane, as well as to NMDA receptors, demonstrating that γ-secretase is present in the postsynaptic plasma membrane. Importantly, the expression of γ-secretase increased in the pre- and postsynaptic compartments with the size of the synapse, suggesting a correlation between γ-secretase activity and synapse maturation. Thus, our data shows the synaptic location with high precision in three dimensions and settles the long-lasting debate on the synaptic location of γ-secretase.
Sigma-Aldrich
Anti-Microtubule-Associated Protein 2 (MAP2) Antibody, Chemicon®, from rabbit