Merck
  • Thymosin beta 15A (TMSB15A) is a predictor of chemotherapy response in triple-negative breast cancer.
Thymosin beta 15A (TMSB15A) is a predictor of chemotherapy response in triple-negative breast cancer.
British journal of cancer (2012-10-20)
S Darb-Esfahani, R Kronenwett, G von Minckwitz, C Denkert, M Gehrmann, A Rody, J Budczies, J C Brase, M K Mehta, H Bojar, B Ataseven, T Karn, E Weiss, D M Zahm, F Khandan, M Dietel, S Loibl
ABSTRACT
Biomarkers predictive of pathological complete response (pCR) to neoadjuvant chemotherapy (NACT) of breast cancer are urgently needed. Using a training/validation approach for detection of predictive biomarkers in HER2-negative breast cancer, pre-therapeutic core biopsies from four independent cohorts were investigated: Gene array data were analysed in fresh frozen samples of two cohorts (n=86 and n=55). Quantitative reverse transcription polymerase chain reaction (qRT-PCR) was performed in formalin-fixed, paraffin-embedded (FFPE) samples from two neoadjuvant phase III trials (GeparTrio, n=212, and GeparQuattro, n=383). A strong predictive capacity of thymosin beta 15 (TMSB15A) gene expression was evident in both fresh frozen cohorts (P<0.0001; P<0.0042). In the GeparTrio FFPE training cohort, a significant linear correlation between TMSB15A expression and pCR was apparent in triple-negative breast cancer (TNBC) (n=61, P=0.040). A cutoff point was then defined that divided TNBC into a low and a high expression group (pCR rate 16.0% vs 47.2%). Both linear correlation of TMSB15A mRNA levels (P=0.017) and the pre-defined cutoff point were validated in 134 TNBC from GeparQuattro (pCR rate 36.8% vs 17.0%, P=0.020). No significant predictive capacity was observed in luminal carcinomas from GeparTrio and GeparQuattro. In TNBC, TMSB15A gene expression analysis might help to select patients with a high chance for pCR after NACT.
Sigma-Aldrich
MISSION® esiRNA, targeting human TMSB15A