Merck
  • Sphingosine kinase 1 serves as a pro-viral factor by regulating viral RNA synthesis and nuclear export of viral ribonucleoprotein complex upon influenza virus infection.
Sphingosine kinase 1 serves as a pro-viral factor by regulating viral RNA synthesis and nuclear export of viral ribonucleoprotein complex upon influenza virus infection.
PloS one (2013-10-19)
Young-Jin Seo, Curtis J Pritzl, Madhuvanthi Vijayan, Kavita Bomb, Mariah E McClain, Stephen Alexander, Bumsuk Hahm
ABSTRACT
Influenza continues to pose a threat to humans by causing significant morbidity and mortality. Thus, it is imperative to investigate mechanisms by which influenza virus manipulates the function of host factors and cellular signal pathways. In this study, we demonstrate that influenza virus increases the expression and activation of sphingosine kinase (SK) 1, which in turn regulates diverse cellular signaling pathways. Inhibition of SK suppressed virus-induced NF-κB activation and markedly reduced the synthesis of viral RNAs and proteins. Further, SK blockade interfered with activation of Ran-binding protein 3 (RanBP3), a cofactor of chromosome region maintenance 1 (CRM1), to inhibit CRM1-mediated nuclear export of the influenza viral ribonucleoprotein complex. In support of this observation, SK inhibition altered the phosphorylation of ERK, p90RSK, and AKT, which is the upstream signal of RanBP3/CRM1 activation. Collectively, these results indicate that SK is a key pro-viral factor regulating multiple cellular signal pathways triggered by influenza virus infection.
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human KCNN1
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)