Merck
Aclidinium inhibits human lung fibroblast to myofibroblast transition.
Thorax (2011-10-01)
Javier Milara, Adela Serrano, Teresa Peiró, Amadeu Gavaldà, Montserrat Miralpeix, Esteban Jesús Morcillo, Julio Cortijo
ABSTRACT
Fibroblast to myofibroblast transition is believed to contribute to airway remodelling in lung diseases such as asthma and chronic obstructive pulmonary disease. This study examines the role of aclidinium, a new long-acting muscarinic antagonist, on human fibroblast to myofibroblast transition. Human bronchial fibroblasts were stimulated with carbachol (10(-8) to 10(-5) M) or transforming growth factor-β1 (TGF-β1; 2 ng/ml) in the presence or absence of aclidinium (10(-9) to 10(-7) M) or different drug modulators for 48 h. Characterisation of myofibroblasts was performed by analysis of collagen type I and α-smooth muscle actin (α-SMA) mRNA and protein expression as well as α-SMA microfilament immunofluorescence. ERK1/2 phosphorylation, RhoA-GTP and muscarinic receptors (M) 1, 2 and 3 protein expression were determined by western blot analysis and adenosine 3'-5' cyclic monophosphate levels were determined by ELISA. Proliferation and migration of fibroblasts were also assessed. Collagen type I and α-SMA mRNA and protein expression, as well as percentage α-SMA microfilament-positive cells, were upregulated in a similar way by carbachol and TGF-β1, and aclidinium reversed these effects. Carbachol-induced myofibroblast transition was mediated by an increase in ERK1/2 phosphorylation, RhoA-GTP activation and cyclic monophosphate downregulation as well as by the autocrine TGF-β1 release, which were effectively reduced by aclidinium. TGF-β1 activated the non-neuronal cholinergic system. Suppression of M1, M2 or M3 partially prevented carbachol- and TGF-β1-induced myofibroblast transition. Aclidinium dose-dependently reduced fibroblast proliferation and migration. Aclidinium inhibits human lung fibroblast to myofibrobast transition.
Sigma-Aldrich
p-Fluorohexahydro-sila-difenidol hydrochloride, powder, ≥98% (HPLC)
Sigma-Aldrich
N6,2′-O-Dibutyryladenosine 3′,5′-cyclic monophosphate sodium salt, ≥97% (HPLC), powder
Sigma-Aldrich
PD 98,059, solid
Sigma-Aldrich
Methoctramine hydrate, ≥97% (NMR), solid
Sigma-Aldrich
Carbamoylcholine chloride, ≥98% (titration), crystalline
Sigma-Aldrich
Y-27632 dihydrochloride, ≥98% (HPLC)