Merck
Regulation of axon regeneration by the RNA repair and splicing pathway.
Nature neuroscience (2015-05-12)
Yuanquan Song, David Sretavan, Ernesto A Salegio, Jim Berg, Xi Huang, Tong Cheng, Xin Xiong, Shan Meltzer, Chun Han, Trong-Tuong Nguyen, Jacqueline C Bresnahan, Michael S Beattie, Lily Yeh Jan, Yuh Nung Jan
ABSTRACT
Mechanisms governing a neuron's regenerative ability are important but not well understood. We identify Rtca (RNA 3'-terminal phosphate cyclase) as an inhibitor of axon regeneration. Removal of Rtca cell-autonomously enhanced axon regrowth in the Drosophila CNS, whereas its overexpression reduced axon regeneration in the periphery. Rtca along with the RNA ligase Rtcb and its catalyst Archease operate in the RNA repair and splicing pathway important for stress-induced mRNA splicing, including that of Xbp1, a cellular stress sensor. Drosophila Rtca and Archease had opposing effects on Xbp1 splicing, and deficiency of Archease or Xbp1 impeded axon regeneration in Drosophila. Moreover, overexpressing mammalian Rtca in cultured rodent neurons reduced axonal complexity in vitro, whereas reducing its function promoted retinal ganglion cell axon regeneration after optic nerve crush in mice. Our study thus links axon regeneration to cellular stress and RNA metabolism, revealing new potential therapeutic targets for treating nervous system trauma.
Sigma-Aldrich
Anti-Tau-1 Antibody, clone PC1C6, clone PC1C6, Chemicon®, from mouse
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse