Merck
  • Imaging cellular structures in super-resolution with SIM, STED and Localisation Microscopy: A practical comparison.
Imaging cellular structures in super-resolution with SIM, STED and Localisation Microscopy: A practical comparison.
Scientific reports (2016-06-07)
Eva Wegel, Antonia Göhler, B Christoffer Lagerholm, Alan Wainman, Stephan Uphoff, Rainer Kaufmann, Ian M Dobbie
ABSTRACT
Many biological questions require fluorescence microscopy with a resolution beyond the diffraction limit of light. Super-resolution methods such as Structured Illumination Microscopy (SIM), STimulated Emission Depletion (STED) microscopy and Single Molecule Localisation Microscopy (SMLM) enable an increase in image resolution beyond the classical diffraction-limit. Here, we compare the individual strengths and weaknesses of each technique by imaging a variety of different subcellular structures in fixed cells. We chose examples ranging from well separated vesicles to densely packed three dimensional filaments. We used quantitative and correlative analyses to assess the performance of SIM, STED and SMLM with the aim of establishing a rough guideline regarding the suitability for typical applications and to highlight pitfalls associated with the different techniques.
Sigma-Aldrich
Schneider′s Insect Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for insect cell culture