Merck
  • Bafilomycin A1 activates HIF-dependent signalling in human colon cancer cells via mitochondrial uncoupling.
Bafilomycin A1 activates HIF-dependent signalling in human colon cancer cells via mitochondrial uncoupling.
Bioscience reports (2012-09-05)
Alexander V Zhdanov, Ruslan I Dmitriev, Dmitri B Papkovsky
ABSTRACT
Mitochondrial uncoupling is implicated in many patho(physiological) states. Using confocal live cell imaging and an optical O2 sensing technique, we show that moderate uncoupling of the mitochondria with plecomacrolide Baf (bafilomycin A1) causes partial depolarization of the mitochondria and deep sustained deoxygenation of human colon cancer HCT116 cells subjected to 6% atmospheric O2. A decrease in iO2 (intracellular O2) to 0-10 μM, induced by Baf, is sufficient for stabilization of HIFs (hypoxia inducible factors) HIF-1α and HIF-2α, coupled with an increased expression of target genes including GLUT1 (glucose transporter 1), HIF PHD2 (prolyl hydroxylase domain 2) and CAIX (carbonic anhydrase IX). Under the same hypoxic conditions, treatment with Baf causes neither decrease in iO2 nor HIF-α stabilization in the low-respiring HCT116 cells deficient in COX (cytochrome c-oxidase). Both cell types display equal capacities for HIF-α stabilization by hypoxia mimetics DMOG (dimethyloxalylglycine) and CoCl2, thus suggesting that the effect of Baf under hypoxia is driven mainly by mitochondrial respiration. Altogether, by activating HIF signalling under moderate hypoxia, mitochondrial uncoupling can play an important regulatory role in colon cancer metabolism and modulate adaptation of cancer cells to natural hypoxic environments.
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Anti-Goat/Sheep IgG−Peroxidase antibody, Mouse monoclonal, clone GT-34, purified from hybridoma cell culture