Merck
  • pGlyco: a pipeline for the identification of intact N-glycopeptides by using HCD- and CID-MS/MS and MS3.
pGlyco: a pipeline for the identification of intact N-glycopeptides by using HCD- and CID-MS/MS and MS3.
Scientific reports (2016-05-04)
Wen-Feng Zeng, Ming-Qi Liu, Yang Zhang, Jian-Qiang Wu, Pan Fang, Chao Peng, Aiying Nie, Guoquan Yan, Weiqian Cao, Chao Liu, Hao Chi, Rui-Xiang Sun, Catherine C L Wong, Si-Min He, Pengyuan Yang
ABSTRACT
Confident characterization of the microheterogeneity of protein glycosylation through identification of intact glycopeptides remains one of the toughest analytical challenges for glycoproteomics. Recently proposed mass spectrometry (MS)-based methods still have some defects such as lack of the false discovery rate (FDR) analysis for the glycan identification and lack of sufficient fragmentation information for the peptide identification. Here we proposed pGlyco, a novel pipeline for the identification of intact glycopeptides by using complementary MS techniques: 1) HCD-MS/MS followed by product-dependent CID-MS/MS was used to provide complementary fragments to identify the glycans, and a novel target-decoy method was developed to estimate the false discovery rate of the glycan identification; 2) data-dependent acquisition of MS3 for some most intense peaks of HCD-MS/MS was used to provide fragments to identify the peptide backbones. By integrating HCD-MS/MS, CID-MS/MS and MS3, intact glycopeptides could be confidently identified. With pGlyco, a standard glycoprotein mixture was analyzed in the Orbitrap Fusion, and 309 non-redundant intact glycopeptides were identified with detailed spectral information of both glycans and peptides.
Sigma-Aldrich
Immunoglobulin G from human serum, ≥95% (GE)
Sigma-Aldrich
IgM from human serum, reagent grade, ~95% (HPLC), buffered aqueous solution