Merck
  • High-throughput proteomics reveal alarmins as amplifiers of tissue pathology and inflammation after spinal cord injury.
High-throughput proteomics reveal alarmins as amplifiers of tissue pathology and inflammation after spinal cord injury.
Scientific reports (2016-02-24)
Athanasios Didangelos, Michele Puglia, Michaela Iberl, Candela Sanchez-Bellot, Bernd Roschitzki, Elizabeth J Bradbury
ABSTRACT
Spinal cord injury is characterized by acute cellular and axonal damage followed by aggressive inflammation and pathological tissue remodelling. The biological mediators underlying these processes are still largely unknown. Here we apply an innovative proteomics approach targeting the enriched extracellular proteome after spinal cord injury for the first time. Proteomics revealed multiple matrix proteins not previously associated with injured spinal tissue, including small proteoglycans involved in cell-matrix adhesion and collagen fibrillogenesis. Network analysis of transcriptomics and proteomics datasets uncovered persistent overexpression of extracellular alarmins that can trigger inflammation via pattern recognition receptors. In mechanistic experiments, inhibition of toll-like receptor-4 (TLR4) and the receptor for advanced glycation end-products (RAGE) revealed the involvement of alarmins in inflammatory gene expression, which was found to be dominated by IL1 and NFκΒ signalling. Extracellular high-mobility group box-1 (HMGB1) was identified as the likely endogenous regulator of IL1 expression after injury. These data reveal a novel tissue remodelling signature and identify endogenous alarmins as amplifiers of the inflammatory response that promotes tissue pathology and impedes neuronal repair after spinal cord injury.
Sigma-Aldrich
IL-1RA murine, recombinant, expressed in E. coli, ≥98% (SDS-PAGE)
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 3, DMSO solution
Sigma-Aldrich
Protease Inhibitor Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
TPCA-1, ≥95% (HPLC)