Merck
  • The Role of Prostaglandins and COX-Enzymes in Chondrogenic Differentiation of ATDC5 Progenitor Cells.
The Role of Prostaglandins and COX-Enzymes in Chondrogenic Differentiation of ATDC5 Progenitor Cells.
PloS one (2016-04-07)
Marjolein M J Caron, Pieter J Emans, Kathleen Sanen, Don A M Surtel, Andy Cremers, Daan Ophelders, Lodewijk W van Rhijn, Tim J M Welting
ABSTRACT
NSAIDs are used to relieve pain and decrease inflammation by inhibition of cyclooxygenase (COX)-catalyzed prostaglandin (PG) synthesis. PGs are fatty acid mediators involved in cartilage homeostasis, however the action of their synthesizing COX-enzymes in cartilage differentiation is not well understood. In this study we hypothesized that COX-1 and COX-2 have differential roles in chondrogenic differentiation. ATDC5 cells were differentiated in the presence of COX-1 (SC-560, Mofezolac) or COX-2 (NS398, Celecoxib) specific inhibitors. Specificity of the NSAIDs and inhibition of specific prostaglandin levels were determined by EIA. Prostaglandins were added during the differentiation process. Chondrogenic outcome was determined by gene- and protein expression analyses. Inhibition of COX-1 prevented Col2a1 and Col10a1 expression. Inhibition of COX-2 resulted in decreased Col10a1 expression, while Col2a1 remained unaffected. To explain this difference expression patterns of both COX-enzymes as well as specific prostaglandin concentrations were determined. Both COX-enzymes are upregulated during late chondrogenic differentiation, whereas only COX-2 is briefly expressed also early in differentiation. PGD2 and PGE2 followed the COX-2 expression pattern, whereas PGF2α and TXA2 levels remained low. Furthermore, COX inhibition resulted in decreased levels of all tested PGs, except for PGD2 and PGF2α in the COX-1 inhibited condition. Addition of PGE2 and PGF2α resulted in increased expression of chondrogenic markers, whereas TXA2 increased expression of hypertrophic markers. Our findings point towards a differential role for COX-enzymes and PG-production in chondrogenic differentiation of ATDC5 cells. Ongoing research is focusing on further elucidating the functional partition of cyclooxygenases and specific prostaglandin production.
Sigma-Aldrich
Anti-Collagen Type X Rabbit pAb, liquid, Calbiochem®