- Bioluminescent bioreporter pad biosensor for monitoring water toxicity.
Bioluminescent bioreporter pad biosensor for monitoring water toxicity.
Talanta (2016-01-01)
Tim Axelrod, Evgeni Eltzov, Robert S Marks
PMID26717844
ABSTRACT
Toxicants in water sources are of concern. We developed a tool that is affordable and easy-to-use for monitoring toxicity in water. It is a biosensor composed of disposable bioreporter pads (calcium alginate matrix with immobilized bacteria) and a non-disposable CMOS photodetector. Various parameters to enhance the sensor's signal have been tested, including the effect of alginate and bacterium concentrations. The effect of various toxicants, as well as, environmental samples were tested by evaluating their effect on bacterial luminescence. This is the first step in the creation of a sensitive and simple operative tool that may be used in different environments.