Merck
  • The Role of Sonic Hedgehog in the Specification of Human Cortical Progenitors In Vitro.
The Role of Sonic Hedgehog in the Specification of Human Cortical Progenitors In Vitro.
Cerebral cortex (New York, N.Y. : 1991) (2014-08-26)
Nevena V Radonjić, Fani Memi, Juan Alberto Ortega, Nicole Glidden, Haiying Zhan, Nada Zecevic
ABSTRACT
Impaired sonic hedgehog (Shh) signaling is involved in the pathology of cortical formation found in neuropsychiatric disorders. However, its role in the specification of human cortical progenitors is not known. Here, we report that Shh is expressed in the human developing cortex at mid-gestation by radial glia cells (RGCs) and cortical neurons. We used RGC cultures, established from the dorsal (cortical) telencephalon of human brain at mid-gestation to study the effect of Shh signaling. Cortical RGCs in vitro maintained their regional characteristics, expressed components of Shh signaling, and differentiated into Nkx2.1, Lhx6, and calretinin-positive (CalR(+)) cells, potential cortical interneuron progenitors. Treatment with exogenous Shh increased the pool of Nkx2.1(+) progenitors, decreased Lhx6 expression, and suppressed the generation of CalR(+) cells. The blockade of endogenous Shh signaling increased the number of CalR(+) cells, but did not affect Nkx2.1 expression, implying the existence of parallel Shh-independent pathways for cortical Nkx2.1 regulation. These results support the idea that, during human brain development, Shh plays an important role in the specification of cortical progenitors. Since direct functional studies in humans are limited, the in vitro system that we established here could be of great interest for modeling the development of human cortical progenitors.
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Calretinin Antibody, clone 6B8.2, clone 6B8.2, Chemicon®, from mouse
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Monoclonal Anti-Parvalbumin antibody produced in mouse, clone PARV-19, ascites fluid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse