Merck
  • Expression of PAX2 and PTEN Correlates to Therapy Response in Endometrial Hyperplasia.
Expression of PAX2 and PTEN Correlates to Therapy Response in Endometrial Hyperplasia.
Anticancer research (2015-12-08)
Anne Ørbo, Marit Arnes, Lena Myreng Lyså, Bjørn Straume
ABSTRACT
To investigate if a levonorgestrel-impregnated intrauterine system (LNG-IUS) was more efficient compared to oral progestin in the clearance of the paired box 2 gene (PAX2) - and phosphatase and tensin homolog (PTEN)-null endometrial glands and assess the significance of PAX2- and PTEN-null glands as markers for therapy response in endometrial hyperplasia. Immunohistochemical staining using antibodies against PAX2 and PTEN was performed in 141 pre- and post-treatment endometrial biopsies comparing the effect of LNG-IUS, 10 mg medroxyprogesterone acetate (MPA) taken continuously, or 10 mg MPA taken 10 days per cycle for six months. PAX2- and PTEN-null glands were investigated by light microscopy in pre-and post-treatment biopsies. Clearance of PAX2- and PTEN-null glands was significantly more efficient by LNG-IUS compared to oral MPA (p<0.000 and p=0.008, respectively) and significantly related to therapy response (p<0.000 and p=0.002, respectively).
Product Description
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea