- Efficient microwave-assisted one-pot preparation of angular 2,2-dimethyl-2H-chromone containing compounds.
Efficient microwave-assisted one-pot preparation of angular 2,2-dimethyl-2H-chromone containing compounds.
Tetrahedron letters (2010-10-12)
Ting Zhou, Qian Shi, Kuo Hsing Lee
PMID20936082
ABSTRACT
A novel and efficient microwave-assisted one-pot reaction was developed to synthesize angular 2,2-dimethyl-2H-chromone containing compounds, which is the first and key step in the synthesis of potent DCK and DCP anti-HIV agents. The newly developed microwave synthesis conditions dramatically shortened the reaction time from 2 days to 4 hours with improved yields.