Merck
  • Novel dual-color immunohistochemical methods for detecting ERG-PTEN and ERG-SPINK1 status in prostate carcinoma.
Novel dual-color immunohistochemical methods for detecting ERG-PTEN and ERG-SPINK1 status in prostate carcinoma.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc (2013-01-26)
Ritu Bhalla, Lakshmi P Kunju, Scott A Tomlins, Kelly Christopherson, Connie Cortez, Shannon Carskadon, Javed Siddiqui, Kyung Park, Juan Miguel Mosquera, Gary A Pestano, Mark A Rubin, Arul M Chinnaiyan, Nallasivam Palanisamy
ABSTRACT
Identification of new molecular markers has led to the molecular classification of prostate cancer based on driving genetic lesions. The translation of these discoveries for clinical use necessitates the development of simple, reliable and rapid detection systems to screen patients for specific molecular aberrations. We developed two dual-color immunohistochemistry-based assays for the simultaneous assessment of ERG-PTEN and ERG-SPINK1 in prostate cancer. A total of 232 cases from 184 localized and 48 metastatic prostate cancers were evaluated for ERG-PTEN and 284 cases from 228 localized and 56 metastatic prostate cancers were evaluated for ERG-SPINK1. Of the 232 cases evaluated for ERG-PTEN, 81 (35%) ERG-positive and 77 (33%) PTEN-deleted cases were identified. Of the 81 ERG-positive cases, PTEN loss was confirmed in 35 (15%) cases by fluorescence in situ hybridization (FISH). PTEN status was concordant in 203 cases (sensitivity 90% and specificity 87%; P<0.0001) by both immunohistochemisty and FISH; however, immunohistochemisty could not distinguish between heterozygous and homozygous deletion status of PTEN. Of the 284 cases evaluated for ERG-SPINK1, 111 (39%) cases were positive for ERG. In the remaining 173 ERG-negative cases, SPINK1 was positive in 26 (9%) cases. SPINK1 expression was found to be mutually exclusive with ERG expression; however, we identified two cases, of which one showed concomitant expression of ERG and SPINK1 in the same tumor foci, and in the second case ERG and SPINK1 were seen in two independent foci of the same tumor nodule. Unlike the homogenous ERG staining in cancer tissues, heterogeneous SPINK1 staining was observed in the majority of the cases. Further studies are required to understand the molecular heterogeneity of cases with concomitant ERG-SPINK1 expression. Automated dual ERG-PTEN and ERG-SPINK1 immunohistochemisty assays are simple, reliable and portable across study sites for the simultaneous assessment of these proteins in prostate cancer.
Product Description
Roche
DIG-Nick Translation Mix, sufficient for 40 labeling reactions, pkg of 160 μL, suitable for hybridization
Roche
Biotin-Nick Translation Mix, suitable for hybridization, solution, sufficient for 40 labeling reactions, pkg of 160 μL