Merck
  • Grp1-associated scaffold protein regulates skin homeostasis after ultraviolet irradiation.
Grp1-associated scaffold protein regulates skin homeostasis after ultraviolet irradiation.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2014-01-11)
Anand Venkataraman, Daniel J Coleman, Daniel J Nevrivy, Tulley Long, Chrissa Kioussi, Arup K Indra, Mark Leid
ABSTRACT
Grp1-associated scaffold protein (Grasp), the product of a retinoic acid-induced gene in P19 embryonal carcinoma cells, is expressed primarily in brain, heart, and lung of the mouse. We report herein that Grasp transcripts are also found in mouse skin in which the Grasp gene is robustly induced following acute ultraviolet-B (UVB) exposure. Grasp(-/-) mice were found to exhibit delayed epidermal proliferation and a blunted apoptotic response after acute UVB exposure. Immunohistochemical analyses revealed that the nuclear residence time of the tumor suppressor protein p53 was reduced in Grasp(-/-) mice after UVB exposure. Taken together, our results suggest that a physiological role of Grasp may be to regulate skin homeostasis after UVB exposure, potentially by influencing p53-mediated apoptotic responses in skin.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-p53 (Ab-3) (Mutant) Mouse mAb (PAb240), liquid, clone PAb240, Calbiochem®