Merck
  • SIRT1 attenuates PAF-induced MMP-2 production via down-regulation of PAF receptor expression in vascular smooth muscle cells.
SIRT1 attenuates PAF-induced MMP-2 production via down-regulation of PAF receptor expression in vascular smooth muscle cells.
Vascular pharmacology (2015-05-15)
Yun H Kim, Jin U Bae, Seung J Lee, So Y Park, Chi D Kim
ABSTRACT
Silent mating type information regulation 2 homolog 1 (SIRT1) is known as a key regulator in the protection of various vascular disorders, however, no direct evidences have been reported in the progression of atherosclerosis. Considering the pivotal role of matrix metalloproteinase-2 (MMP-2) in plaque destabilization, this study investigated the role of SIRT1 on MMP-2 production in vascular smooth muscle cells (VSMCs) induced by platelet activating factor (PAF, 1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine). In VSMCs stimulated with resveratrol, SIRT1 activator, PAF receptor (PAFR) was internalized and then its protein levels were diminished. It was attenuated in cells pretreated with proteasome or lysosome inhibitor. Also, the degradation of PAFR in SIRT1-stimulated cells was significantly attenuated by β-arrestin2 depletion. In cells treated with nicotinamide, SIRT1 deacetylase inhibitor, PAFR internalization by resveratrol or reSIRT1 was inhibited, demonstrating that deacetylation of SIRT1 is an important step in SIRT1-induced PAFR down-regulation. Moreover, PAF-induced MMP-2 production in VSMCs and aorta was attenuated by resveratrol. In the aorta of SIRT1 transgenic mice, the PAF-induced MMP-2 expression was prominently attenuated compared to that in wild type mice. Taken together, it was suggested that SIRT1 down-regulated PAFR in VSMCs via β-arrestin2-mediated internalization and degradation, leading to an inhibition of PAF-induced MMP-2 production.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Sirtuin 1 human, recombinant, expressed in E. coli, ≥60% (SDS-PAGE)
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
SIRT1 human, recombinant, expressed in E. coli, N-terminal histidine tagged, ≥90% (SDS-PAGE), buffered aqueous glycerol solution