Merck
  • Insights into the role of immunosenescence during varicella zoster virus infection (shingles) in the aging cell model.
Insights into the role of immunosenescence during varicella zoster virus infection (shingles) in the aging cell model.
Oncotarget (2015-10-17)
Ji-Ae Kim, Seul-Ki Park, Mukesh Kumar, Chan-Hee Lee, Ok Sarah Shin
ABSTRACT
Varicella zoster virus (VZV) is the etiological agent of shingles, a painful skin rash that affects a significant proportion of the elderly population. In the present study, we used two aging cell models, Hutchinson-Gilford progeria syndrome (HGPS) fibroblasts and stress or replicative senescence-induced normal human dermal fibroblasts (NHDFs), to investigate age-associated susceptibility to VZV infection. VZV infectivity titers were significantly associated with donor age in HGPS fibroblasts and senescence induction in NHDFs. High throughput RNA-sequencing (RNA-seq) analysis was performed to assess global and dynamic changes in the host transcriptomes of VZV-infected aging cells. Analysis of differentially expressed genes (DEGs) indicated that VZV infection in aged HGPS fibroblasts resembled that in senescent NHDFs, particularly in terms of genes associated with pattern recognition receptors in virus sensing network, providing novel insights into the mechanisms of senescence-associated susceptibility to VZV infection. Additionally, we identified stimulator of interferon genes (STING) as a potential VZV sensing receptor. Knockdown of STING expression resulted in increased viral replication in primary fibroblasts, whereas STING overexpression led to suppression of VZV plaque formation. In conclusion, our findings highlight the important role of immunosenescence following VZV infection and provide significant insights into the mechanisms underlying cellular sensing of VZV infection and the induction of immune responses in aged skin cells.
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
MISSION® esiRNA, targeting human TMEM173
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tmem173
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
2-Propanol, BioReagent, for molecular biology, ≥99.5%
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
SAFC
L-Glutamine
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)