Merck
  • Hair concentration of essential trace elements in adult non-exposed Russian population.
Hair concentration of essential trace elements in adult non-exposed Russian population.
Environmental monitoring and assessment (2015-10-09)
Anatoly V Skalny, Margarita G Skalnaya, Alexey A Tinkov, Eugeny P Serebryansky, Vasily A Demidov, Yulia N Lobanova, Andrei R Grabeklis, Elena S Berezkina, Irina V Gryazeva, Andrey A Skalny, Oksana A Skalnaya, Nikolay G Zhivaev, Alexandr A Nikonorov
ABSTRACT
Appropriate reference values of hair trace element content are required for correct interpretation of biomonitoring data. The primary objective of the current study was to estimate the reference values of selected essential trace elements in hair of adult Russian population. Involved in current investigation were 7256 occupationally non-exposed adults aged from 20 to 60 years and living in the European part of Russia. Occipital hair essential metal and metalloid (Co, Cr, Cu, Fe, Mn, Se, V, Zn) content was estimated using inductively coupled plasma mass spectrometry. The reference ranges were calculated in accordance with the International Union of Pure and Applied Chemistry (IUPAC) recommendations. Women were characterized by 55, 18, 58, and 7% higher values of hair Co, Cu, Mn, and Zn content as compared to the values observed in men. At the same time, hair Cr, Fe, Se, and V concentration in men significantly exceeded the respective female values by 65, 13, 20, and 56%. Consequently, the reference ranges of essential hair trace elements content should be separately calculated for both men and women. The obtained reference ranges for hair Co, Cr, Cu, Fe, Mn, Se, V, and Zn in men were 0.11-0.67, 0.007-0.045, 10.4-22.6, 11.1-40.5, 0.24-1.05, 0.089-0.480, 0.014-0.083, and 125.7-262.8 μg/g, respectively. The respective values estimated for women were 0.06-0.40, 0.011-0.085, 12.1-44.5, 8.9-25.6, 0.32-2.05, 0.094-0.504, 0.010-0.056, and 140.0-315.1 μg/g. The reference ranges for hair Co (0.07-0.50), Cr (0.009-0.073), Cu (11.8-29.2), Fe (9.6-31.5), Mn (0.29-1.76), Se (0.093-0.482), V (0.011-0.069), and Zn (134.7-301.9) content (μg/g) in the general cohort were also calculated.
Product Description
Yttrium, foil, not light tested, 25x25mm, thickness 0.025mm, as rolled, 99%
Yttrium, foil, not light tested, 50x50mm, thickness 0.025mm, as rolled, 99%
Yttrium, rod, 100mm, diameter 12.5mm, cast, 99%
Yttrium, foil, not light tested, 25x25mm, thickness 0.005mm, as rolled, 99%
Sigma-Aldrich
Yttrium sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.9% trace metals basis
Yttrium, rod, 50mm, diameter 6.35mm, cast, 99%
Yttrium, wire reel, 0.2m, diameter 0.5mm, 99.9%
Yttrium, rod, 100mm, diameter 6.35mm, cast, 99%
Yttrium, wire reel, 0.2m, diameter 1.0mm, 99.9%
Yttrium, rod, 50mm, diameter 12.5mm, cast, 99%
Yttrium, wire reel, 0.1m, diameter 1.0mm, 99.9%
Yttrium, rod, 50mm, diameter 2.0mm, cast, 99%
Yttrium, wire reel, 0.05m, diameter 0.5mm, 99.9%
Yttrium, wire reel, 0.05m, diameter 1.0mm, 99.9%
Yttrium, wire reel, 0.1m, diameter 0.5mm, 99.9%
Sigma-Aldrich
1-Butanol, ACS reagent, ≥99.4%
Sigma-Aldrich
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Yttrium, chips, 99.9% trace rare earth metals basis
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
Sigma-Aldrich
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
1-Butanol, 99.9%
Sigma-Aldrich
Tetramethylammonium hydroxide solution, ACS reagent
Sigma-Aldrich
1-Butanol, BioRenewable, ACS reagent, ≥99.4%
Yttrium, foil, 25mm disks, thickness 0.50mm, as rolled, 99%
Yttrium, foil, not light tested, 100x100mm, thickness 0.025mm, as rolled, 99%
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis