Merck
  • Matching in Vitro Bioaccessibility of Polyphenols and Antioxidant Capacity of Soluble Coffee by Boosted Regression Trees.
Matching in Vitro Bioaccessibility of Polyphenols and Antioxidant Capacity of Soluble Coffee by Boosted Regression Trees.
Journal of agricultural and food chemistry (2015-10-13)
Natalia S Podio, Rebeca López-Froilán, Esther Ramirez-Moreno, Lidwina Bertrand, María V Baroni, María L Pérez-Rodríguez, María-Cortes Sánchez-Mata, Daniel A Wunderlin
ABSTRACT
The aim of this study was to evaluate changes in polyphenol profile and antioxidant capacity of five soluble coffees throughout a simulated gastro-intestinal digestion, including absorption through a dialysis membrane. Our results demonstrate that both polyphenol content and antioxidant capacity were characteristic for each type of studied coffee, showing a drop after dialysis. Twenty-seven compounds were identified in coffee by HPLC-MS, while only 14 of them were found after dialysis. Green+roasted coffee blend and chicory+coffee blend showed the highest and lowest content of polyphenols and antioxidant capacity before in vitro digestion and after dialysis, respectively. Canonical correlation analysis showed significant correlation between the antioxidant capacity and the polyphenol profile before digestion and after dialysis. Furthermore, boosted regression trees analysis (BRT) showed that only four polyphenol compounds (5-p-coumaroylquinic acid, quinic acid, coumaroyl tryptophan conjugated, and 5-O-caffeoylquinic acid) appear to be the most relevant to explain the antioxidant capacity after dialysis, these compounds being the most bioaccessible after dialysis. To our knowledge, this is the first report matching the antioxidant capacity of foods with the polyphenol profile by BRT, which opens an interesting method of analysis for future reports on the antioxidant capacity of foods.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%
Sigma-Aldrich
D-(−)-Quinic acid, 98%
Sigma-Aldrich
p-Coumaric acid, ≥98.0% (HPLC)
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, for spectrophotometric det. (of Fe), ≥98%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%
Sigma-Aldrich
Formic acid, reagent grade, ≥95%
Sigma-Aldrich
Formic acid, ACS reagent, ≥96%
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.