Merck
  • Biogenic volatile organic compound analyses by PTR-TOF-MS: Calibration, humidity effect and reduced electric field dependency.
Biogenic volatile organic compound analyses by PTR-TOF-MS: Calibration, humidity effect and reduced electric field dependency.
Journal of environmental sciences (China) (2015-06-05)
Xiaobing Pang
ABSTRACT
Green leaf volatiles (GLVs) emitted by plants after stress or damage induction are a major part of biogenic volatile organic compounds (BVOCs). Proton transfer reaction time-of-flight mass spectrometry (PTR-TOF-MS) is a high-resolution and sensitive technique for in situ GLV analyses, while its performance is dramatically influenced by humidity, electric field, etc. In this study the influence of gas humidity and the effect of reduced field (E/N) were examined in addition to measuring calibration curves for the GLVs. Calibration curves measured for seven of the GLVs in dry air were linear, with sensitivities ranging from 5 to 10 ncps/ppbv (normalized counts per second/parts per billion by volume). The sensitivities for most GLV analyses were found to increase by between 20% and 35% when the humidity of the sample gas was raised from 0% to 70% relative humidity (RH) at 21°C, with the exception of (E)-2-hexenol. Product ion branching ratios were also affected by humidity, with the relative abundance of the protonated molecular ions and higher mass fragment ions increasing with humidity. The effect of reduced field (E/N) on the fragmentation of GLVs was examined in the drift tube of the PTR-TOF-MS. The structurally similar GLVs are acutely susceptible to fragmentation following ionization and the fragmentation patterns are highly dependent on E/N. Overall the measured fragmentation patterns contain sufficient information to permit at least partial separation and identification of the isomeric GLVs by looking at differences in their fragmentation patterns at high and low E/N.
Sigma-Aldrich
cis-3-Hexen-1-ol, natural, >98%, FCC, FG
Sigma-Aldrich
Hexanal, 98%
Sigma-Aldrich
cis-3-Hexen-1-ol, 98%
Sigma-Aldrich
1-Hexanol, anhydrous, ≥99%
Sigma-Aldrich
Hexyl alcohol, natural, ≥98%, FCC, FG
Sigma-Aldrich
Hexyl alcohol, FCC, FG
Sigma-Aldrich
trans-2-Hexen-1-al, ≥95%, FCC, FG
Sigma-Aldrich
trans-2-Hexen-1-ol, ≥95%, FCC, FG
Sigma-Aldrich
Hexanal, natural, ≥95%, FG
Sigma-Aldrich
1-Hexanol, ReagentPlus®, ≥99.5% (GC)
Sigma-Aldrich
Hexanal, ≥97%, FCC, FG
Sigma-Aldrich
trans-2-Hexen-1-ol, 96%
Sigma-Aldrich
trans-2-Hexen-1-al, 98%
Sigma-Aldrich
cis-3-Hexenyl acetate, ≥98%, stabilized, FCC, FG
Sigma-Aldrich
Hexyl acetate, 99%
Sigma-Aldrich
Hexyl acetate, natural, ≥98%, FCC, FG
Sigma-Aldrich
Hexyl acetate, ≥98%, FCC, FG
Sigma-Aldrich
trans-3-Hexen-1-ol, ≥95%, stabilized
Sigma-Aldrich
trans-3-Hexen-1-ol, 97%