Merck
  • Bioavailability of Pyrene Associated with Suspended Sediment of Different Grain Sizes to Daphnia magna as Investigated by Passive Dosing Devices.
Bioavailability of Pyrene Associated with Suspended Sediment of Different Grain Sizes to Daphnia magna as Investigated by Passive Dosing Devices.
Environmental science & technology (2015-07-23)
Xiaotian Zhang, Xinghui Xia, Husheng Li, Baotong Zhu, Jianwei Dong
ABSTRACT
Suspended sediment (SPS) is widely present in rivers around the world. However, the bioavailability of hydrophobic organic compounds (HOCs) associated with SPS is not well understood. In this work, the influence of SPS grain size on the bioavailability of SPS-associated pyrene to Daphnia magna was studied using a passive dosing device, which maintained a constant freely dissolved pyrene concentration (Cfree) in the exposure systems. The immobilization and protein as well as enzymatic activities of Daphnia magna were investigated to study the bioavailability of SPS-associated pyrene. With Cfree of pyrene ranging from 20.0 to 60.0 μg L(-1), the immobilization of Daphnia magna in the presence of 1 g L(-1) SPS was 1.11-2.89 times that in the absence of SPS. The immobilization caused by pyrene associated with different grain size SPS was on the order of 50-100 μm > 0-50 μm > 100-150 μm. When pyrene Cfree was 20.0 μg L(-1), the immobilization caused by pyrene associated with 50-100 μm SPS was 1.42 and 2.43 times that with 0-50 and 100-150 μm SPS, respectively. The protein and enzymatic activities of Daphnia magna also varied with the SPS grain size. The effect of SPS grain size on the bioavailability of SPS-associated pyrene was mainly due to the difference in SPS ingestion by Daphnia magna and SPS composition, especially the organic carbon type, among the three size fractions. This study suggests that not only the concentration but also the size distribution of SPS should be considered for the development of a biological effect database and establishment of water quality criteria for HOCs in natural waters.
Sigma-Aldrich
m-Terphenyl, 99%
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Pyrene, sublimed grade, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Poly(dimethylsiloxane), viscosity 1.0 cSt (25 °C)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Hexamethyldisiloxane, viscosity 0.65 cSt (25 °C)
Sigma-Aldrich
Pyrene, 98%
Sigma-Aldrich
Pyrene, puriss. p.a., ≥99.0% (GC)
Sigma-Aldrich
Methanol
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Hexane, anhydrous, 95%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.