Merck
  • Structure and functions of the placenta in common minke (Balaenoptera acutorostrata), Bryde's (B. brydei) and sei (B. borealis) whales.
Structure and functions of the placenta in common minke (Balaenoptera acutorostrata), Bryde's (B. brydei) and sei (B. borealis) whales.
The Journal of reproduction and development (2015-06-23)
Chiyo Kitayama, Motoki Sasaki, Hajime Ishikawa, Toshihiro Mogoe, Seiji Ohsumi, Yutaka Fukui, Teguh Budipitojo, Daisuke Kondoh, Nobuo Kitamura
ABSTRACT
The structure and functions of placentas were examined in 3 species of rorqual whales, common minke (Balaenoptera acutorostrata), Bryde's (B. brydei) and sei (B. borealis) whales, with the aim of confirming the structural characteristics of the chorion, including the presence of the areolar part, and clarifying steroidogenic activities and fetomaternal interactions in the placentas of these whales. Placentas were collected from the second phase of the Japanese Whale Research Program under Special Permit in the North Pacific (JARPN II). Histological and ultrastructural examinations revealed that these whale placentas were epitheliochorial placentas with the interdigitation of chorionic villi lined by monolayer uninucleate cells (trophoblast cells) and endometrial crypts as well as folded placentation by fold-like chorionic villi. Moreover, well-developed pouch-like areolae were observed in the placentas, and active absorption was suggested in the chorionic epithelial cells of the areolar part (areolar trophoblast cells). Berlin blue staining showed the presence of ferric ions (Fe(3+)) in the uterine glandular epithelial cells and within the stroma of chorionic villi in the areolar part. An immunohistochemical examination revealed tartrate-resistant acid phosphatase (TRAP; known as uteroferrin in uteri) in the cytoplasm of glandular cells and areolar trophoblast cells. This result suggested that, in cetaceans, uteroferrin is used to supply iron to the fetus. Furthermore, immunoreactivity for P450scc and P450arom was detected in trophoblast cells, but not in areolar trophoblast cells, suggesting that trophoblast cells synthesize estrogen in whale placentas. Therefore, we herein immunohistochemically revealed the localization of aromatase and uteroferrin in cetacean placentas during pregnancy for the first time.
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
Osmium tetroxide, Sealed ampule.
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Methanol, NMR reference standard
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Osmium tetroxide, ACS reagent, ≥98.0%
Sigma-Aldrich
Osmium tetroxide, ReagentPlus®, 99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Osmium tetroxide solution, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Osmium tetroxide solution, 2.5 wt. % in tert-butanol