Merck
  • Magnesium and selected parameters of the non-enzymatic antioxidant and immune systems and oxidative stress intensity in the seminal plasma of fertile males.
Magnesium and selected parameters of the non-enzymatic antioxidant and immune systems and oxidative stress intensity in the seminal plasma of fertile males.
Magnesium research (2015-05-15)
Aleksandra Kasperczyk, Michał Dobrakowski, Jolanta Zalejska-Fiolka, Stanisław Horak, Ewa Birkner
ABSTRACT
The present study investigated associations between environmental exposure to magnesium (Mg) and the levels of oxidative stress parameters and selected cytokines, and the antioxidant defense system in the seminal plasma of fertile males. The study population consisted of 57 healthy, non-smoking, fertile men from the southern region of Poland. Based on the median magnesium levels in seminal plasma, subjects were divided into two groups: those with low (Mg-L) and high (Mg-H) magnesium concentrations. Differences were not observed between the Mg-L and Mg-H groups as regards semen volume, pH, count, motility, or morphology of sperm cells. In the Mg-H group, cholesterol levels were significantly higher (77%) compared with the Mg-L group: however, γ-glutamyltranspeptidase activity was significantly lower (21%). The values of total oxidant status were significantly different between the two groups, with 78% higher values observed in the Mg-H group compared with the Mg-L group, whereas malondialdehyde levels did not differ significantly. The values for total antioxidant capacity and uric acid levels were significantly lower in the Mg-H group compared with the Mg-L group (14% and 17%, respectively). However, levels of bilirubin, albumin, thiol groups and α-tocopherol were significantly higher in the Mg-H group (71%, 44%, 35% and 47%, respectively). No associations between Mg levels in the seminal plasma of fertile males and standard semen parameters were found. However, Mg levels may be associated with altered function of the non-enzymatic antioxidant system.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
1-Butanol, anhydrous, 99.8%
Sigma-Aldrich
Quinidine sulfate salt dihydrate
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
1-Butanol, for molecular biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Butyl alcohol, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Cesium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Cesium chloride, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥98%
Sigma-Aldrich
Cesium chloride, Grade I, ≥99.0%
Sigma-Aldrich
Cesium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Cesium chloride, 99.99% trace metals basis
Sigma-Aldrich
Cesium chloride, for molecular biology, ≥99% (silver nitrate titration)
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis
Sigma-Aldrich
Cesium chloride, Grade II, ≥98%
Sigma-Aldrich
Cesium chloride, ≥99.999% trace metals basis