Merck
  • Photoisomerization of Trans Ortho-, Meta-, Para-Nitro Diarylbutadienes: A Case of Regioselectivity.
Photoisomerization of Trans Ortho-, Meta-, Para-Nitro Diarylbutadienes: A Case of Regioselectivity.
Photochemistry and photobiology (2015-08-08)
Harsha Agnihotri, Mahalingavelar Paramasivam, Veerabhadraiah Palakollu, Sriram Kanvah
ABSTRACT
A series of ortho-, meta- and para-substituted trans-nitro aryl (phenyl and pyridyl) butadienes have been synthesized and characterized. The effect of substitution and positional selectivity on their fluorescence and photoisomerization were systematically investigated. Among all dienes, meta- and para-nitro phenyl-substituted derivatives exhibit remarkable solvatochromic emission shifts due to intramolecular charge transfer. On the other hand, ortho derivatives undergo regioselective isomerization upon photoexcitation in contrast to inefficient isomerization of para and meta nitro-substituted dienes. Single crystal X-ray analysis revealed existence of intramolecular hydrogen bonding between the nitro group and the hydrogen of the proximal double bond. This restricts the rotation of the proximal double bond thereby allowing regioselective isomerization. The observations were also supported by NMR spectroscopic studies.
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
Chloroform-d, 99.8 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
Chloroform-d, "100%", 99.95 atom % D
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Chloroform-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%