Merck
  • Relationship between the A(8002)G intronic polymorphism of pre-pro-endothelin-1 gene and the endothelin-1 concentration among Tunisian coronary patients.
Relationship between the A(8002)G intronic polymorphism of pre-pro-endothelin-1 gene and the endothelin-1 concentration among Tunisian coronary patients.
BMC cardiovascular disorders (2015-11-18)
Abdelkader Chalghoum, Yosri Noichri, Azza Dandana, Sana Azaiez, Bruno Baudin, Gouider Jeridi, Abdelhédi Miled, Salima Ferchichi
ABSTRACT
Acute coronary syndromes (ACS) are complex and polygenic diseases which are a real problem of public health. These syndromes require multidisciplinary studies to understand the pathogenesis mechanisms. Our study aims to evaluate the endothelin-1 (ET-1) serum concentration in Tunisian coronary compared to controls healthy, as well as the study of the impact of an intronic polymorphism A (8002) G of pre-pro-endothelin-1 Gene (inactive precursor of ET-1) on the change in serum endothelin-1 and in the susceptibility to Acute coronary syndrome (SCA). Our samples were subdivided into coronary patients (157) and healthy subjects (142). The quantification of the ET-1 concentration was performed by high performance liquid chromatography, the identification of the different genotypes of the polymorphism A(8002)G was made by PCR-RFLP. The association between the ET-1 concentration and identified genotypes was realized by adapted software for descriptive statistics, Statistical Package for the Sociological Sciences (SPSS v 21.0). Our study showed that the concentration of ET-1 was significantly higher in patients compared to controls and that the mutated allele prevails in patients F (G) = 0.78 and there is a minority in controls F (G) = 0.3. Secondly the homozygous genotype GG is associated with higher concentrations of ET-1 in patients and controls, heterozygous genotype AG is associated with intermediaries' values and AA genotype is related to lower values. Although the polymorphism studied is an intronic polymorphism, it is involved in the change in serum concentration of ET-1 and is a candidate gene in susceptibility to SCA. Cardiovascular diseases are "polygenic" pathology and do not obey of the law for transmission of Mendel.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Endothelin 1, ≥97% (HPLC), powder
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~0.025 M in H2O
Sigma-Aldrich
Magnesium chloride solution, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, 2 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm