Merck
  • Synthesis, characterization, antioxidant activity and neuroprotective effects of selenium polysaccharide from Radix hedysari.
Synthesis, characterization, antioxidant activity and neuroprotective effects of selenium polysaccharide from Radix hedysari.
Carbohydrate polymers (2015-04-11)
Dongfeng Wei, Tong Chen, Mingfei Yan, Wanghong Zhao, Fei Li, Weidong Cheng, Lixia Yuan
ABSTRACT
Beta-amyloid (Aβ) peptide, the hallmark of Alzheimer's disease, invokes oxidative damage to neurons and eventually leads to neuronal death. Selenylation modification of polysaccharide obtained from Radix hedysari (RHP) was studied to access antioxidant activities and neuroprotective effects against oxidative stress and apoptosis induced by Aβ25-35 in vitro. A series of the selenylation derivatives of RHP (Se-RHP) was synthesized using nitric acid-sodium selenite (HNO3-Na2SeO3) method. The organic selenium content of Se-RHP increased from 1.04 to 3.29 mg/g. However, compared with the weight-average molecular mass (Mw) of RHP, Mw of Se-RHP showed a significant decrease, and varied from 27.7 kDa to 62.7 kDa. FT-IR spectra and (13)C NMR spectra indicated the selenite groups had been introduced mainly at the C-6 positions of RHP. Compared with RHP, Se-RHP showed greater antioxidant activities in vitro. Furthermore, both RHP and Se-RHP3 had neuroprotective effects against Aβ25-35-induced oxidative stress and apoptosis in SH-SY5Y human neuroblastoma cells, which might be a potential therapeutic agent for preventing or treating neurodegenerative diseases.
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Guaiacol, natural, ≥99%, FG
Sigma-Aldrich
3-tert-Butyl-4-hydroxyanisole, ≥98% (sum of isomers, GC), ≤10% 2-BHA basis (GC)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Nitrotetrazolium Blue chloride, powder, electrophoresis grade
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Guaiacol, oxidation indicator
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Nitrotetrazolium Blue chloride, ≥90.0% (HPLC)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
2-Deoxy-D-ribose, 97% (GC)