Merck
  • Development of an enzymatic assay system of D-lactate using D-lactate dehydrogenase and a UV-LED fluorescent spectrometer.
Development of an enzymatic assay system of D-lactate using D-lactate dehydrogenase and a UV-LED fluorescent spectrometer.
Journal of pharmaceutical and biomedical analysis (2015-08-13)
Chien-Ming Chen, Shih-Ming Chen, Po-Jen Chien, Han-Yin Yu
ABSTRACT
In this study, we aimed to develop a new enzymatic assay system of d-lactate with good precision, accuracy, and sensitivity for the determination of D-lactate concentrations in rat serum. D-Lactate dehydrogenase (D-LDH) was utilized to catalyze D-lactate and NAD(+) to pyruvate and NADH, respectively. The generated NADH was excited by using a 340-nm UV-light-emitting diode (LED), and the fluorescence at 491 nm was detected to determine the concentration of D-lactate in rat serum. The optics, consisting of the sample cuvette, were set on three-dimensional stages to receive the most intensive fluorescence signal into the spectrometer. The optimal conditions of the D-LDH reaction were pH 8.5 and 25 °C for 90 min. The results showed that the new D-lactate assay system had good linearity (R(2)=0.9964) in the calibration range from 5 to 150 μM. Intra-day and inter-day accuracies were in the range of 103.96-109.09% and 102.84-104.59%, respectively, and the intra-day and inter-day precision was 4.28-6.82% and 4.04-12.40%, respectively. Finally, serum D-lactate concentrations determined by the proposed enzymatic assay system were compared with those obtained by a conventional HPLC method. The newly developed D-lactate assay system could detect 10-15 samples in 90 min, whereas the HPLC method could detect only one sample over the same time period.
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Triphenylphosphine, polymer-bound, 100-200 mesh, extent of labeling: ~1-1.5 mmol/g Capacity (Phosphor)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
2,2′-Dithiodipyridine, powder
Sigma-Aldrich
Lithium L-lactate, ≥98% (titration)
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Aldrithiol-2, 98%
Sigma-Aldrich
Triphenylphosphine, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphine, ≥95.0% (GC)
Sigma-Aldrich
Citric acid monohydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., buffer substance, 99.5-102%
Sigma-Aldrich
Citric acid monohydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, E330, 99.5-100.5% (based on anhydrous substance), grit
Sigma-Aldrich
Citric acid monohydrate, reagent grade, ≥98% (GC/titration)
Sigma-Aldrich
Citric acid monohydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Citric acid monohydrate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)