Merck
Anaerobic biosynthesis of the lower ligand of vitamin B12.
Proceedings of the National Academy of Sciences of the United States of America (2015-08-08)
Amrita B Hazra, Andrew W Han, Angad P Mehta, Kenny C Mok, Vadim Osadchiy, Tadhg P Begley, Michiko E Taga
ABSTRACT
Vitamin B12 (cobalamin) is required by humans and other organisms for diverse metabolic processes, although only a subset of prokaryotes is capable of synthesizing B12 and other cobamide cofactors. The complete aerobic and anaerobic pathways for the de novo biosynthesis of B12 are known, with the exception of the steps leading to the anaerobic biosynthesis of the lower ligand, 5,6-dimethylbenzimidazole (DMB). Here, we report the identification and characterization of the complete pathway for anaerobic DMB biosynthesis. This pathway, identified in the obligate anaerobic bacterium Eubacterium limosum, is composed of five previously uncharacterized genes, bzaABCDE, that together direct DMB production when expressed in anaerobically cultured Escherichia coli. Expression of different combinations of the bza genes revealed that 5-hydroxybenzimidazole, 5-methoxybenzimidazole, and 5-methoxy-6-methylbenzimidazole, all of which are lower ligands of cobamides produced by other organisms, are intermediates in the pathway. The bza gene content of several bacterial and archaeal genomes is consistent with experimentally determined structures of the benzimidazoles produced by these organisms, indicating that these genes can be used to predict cobamide structure. The identification of the bza genes thus represents the last remaining unknown component of the biosynthetic pathway for not only B12 itself, but also for three other cobamide lower ligands whose biosynthesis was previously unknown. Given the importance of cobamides in environmental, industrial, and human-associated microbial metabolism, the ability to predict cobamide structure may lead to an improved ability to understand and manipulate microbial metabolism.
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
Sigma-Aldrich
Dicyanocobinamide, ≥93%
Sigma-Aldrich
Cyanocobalamin, meets USP testing specifications
Sigma-Aldrich
Ethanolamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Pyrimidine, ≥98.0%
Sigma-Aldrich
Ethanolamine, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethanolamine, ≥98%
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ethanolamine, liquid, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
2-Aminoimidazole, AldrichCPR
Sigma-Aldrich
Ethanolamine, ≥99%