Merck
  • Probing structural features of self-assembled violanthrone-79 using two dimensional infrared spectroscopy.
Probing structural features of self-assembled violanthrone-79 using two dimensional infrared spectroscopy.
The Journal of chemical physics (2015-06-08)
Jenée D Cyran, Amber T Krummel
ABSTRACT
Two-dimensional infrared (2D IR) spectroscopy was used to characterize the structure of a self-assembled polycyclic aromatic hydrocarbon (PAH), violanthrone-79. A local mode basis was constructed using spectroscopic and computational results of anthrone and monomer violanthrone-79. The vibrational modes in the spectral region 1550-1700 cm(-1), carbonyl stretching and in-plane ring breathing, are used as vibrational probes. The local mode basis and an electrostatic coupling model were applied to three nanoaggregate structures: parallel, antiparallel, and a chiral configuration produced by a 28° rotation from parallel. Angular disorder within each nanoaggregate configuration was also explored. This investigation is a first approach to probe self-assembled PAHs with 2D IR spectroscopy. The experimental and calculated 2D IR spectra align best when the violanthrone-79 molecules are in an anti-parallel configuration within the nanoaggregate.
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Anthrone, ACS reagent, 97%