Merck
  • Disposition of bisphenol AF, a bisphenol A analogue, in hepatocytes in vitro and in male and female Harlan Sprague-Dawley rats and B6C3F1/N mice following oral and intravenous administration.
Disposition of bisphenol AF, a bisphenol A analogue, in hepatocytes in vitro and in male and female Harlan Sprague-Dawley rats and B6C3F1/N mice following oral and intravenous administration.
Xenobiotica; the fate of foreign compounds in biological systems (2015-04-30)
Suramya Waidyanatha, James M Mathews, Purvi R Patel, Sherry R Black, Rodney W Snyder, Timothy R Fennell
ABSTRACT
1. Bisphenol AF (BPAF) is used as a crosslinking agent for polymers and is being considered as a replacement for bisphenol A (BPA). 2. In this study, comparative clearance and metabolism of BPAF and BPA in hepatocytes and the disposition and metabolism of BPAF in rodents following oral administration of 3.4, 34 or 340 mg/kg [(14)C]BPAF were investigated. 3. BPAF was cleared more slowly than BPA in hepatocytes with the rate: rat > mouse > human. 4. [(14)C]BPAF was excreted primarily in feces by 72 h after oral administration to rats (65-80%) and mice (63-72%). Females excreted more in urine (rat, 15%; mouse, 24%) than males (rat, 1-4%; mouse, 10%). Residual tissue radioactivity was <2% of the dose at 72 h. Similar results were observed following intravenous administration. 5. In male rats, 52% of a 340 mg/kg oral dose was excreted in 24 h bile and was mostly comprised of BPAF glucuronide. However, >94% of fecal radioactivity was present as BPAF, suggesting extensive deconjugation in the intestine. 6. Metabolites identified in bile were BPAF-glucuronide, -diglucuronide, -glucuronide sulfate and -sulfate. 7. In conclusion, BPAF was well absorbed following gavage administration and highly metabolized and excreted mostly in the feces as BPAF.
Sigma-Aldrich
Sulfatase from Aerobacter aerogenes, Type VI, buffered aqueous glycerol solution, 2-5 units/mg protein (biuret), 10-20 units/mL
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acylase I from porcine kidney, Grade I, lyophilized powder, ≥1500 units/mg protein
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
β-Glucuronidase from Escherichia coli, Type VII-A, lyophilized powder, 5,000,000-20,000,000 units/g protein, pH 6.8 (30 min assay)