Merck
  • The VGF-Derived Neuropeptide TLQP-21 Shows No Impact on Hormone Secretion in the Isolated Perfused Rat Pancreas.
The VGF-Derived Neuropeptide TLQP-21 Shows No Impact on Hormone Secretion in the Isolated Perfused Rat Pancreas.
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme (2015-01-21)
C B Christiansen, B Svendsen, J J Holst
ABSTRACT
TLQP-21 is a VGF-derived neuropeptide proposed to be involved in regulation of metabolism. More specifically it has been suggested that TLQP-21 has the ability to enhance glucose stimulated insulin secretion, making it a candidate for treatment of patients with type 2 diabetes.In this study, we investigated the impact of TLQP-21 on insulin, glucagon, and somatostatin secretion in the perfused rat pancreas. We found that administration of 5 and 50 nM TLQP-21 had no impact on pancreatic hormone secretion at 3.5 or 8 mM glucose levels. Increasing TLQP-21 (200 nM) and glucose concentration (3.5 and 16 mM) led to a nonsignificant decrease in glucagon secretion, though insulin and somatostatin secretory patterns remained unaffected. In a final set of experiments, perfusions were performed with infusion of 50 and 1 000 nM TLQP-21 to ensure sufficient stimulation. However, administration of TLQP-21 under this setup showed no impact on the pancreatic hormone secretion either. In conclusion, the outcome of this study does not concur with previous findings, suggesting that the effect of TLQP-21 does not directly involve silent hormone secretion.
Sigma-Aldrich
Glucose solution, BioUltra, ~20% in H2O
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
Sigma-Aldrich
L-Arginine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Arginine monohydrochloride, not synthetic, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginine monohydrochloride, reagent grade, ≥98% (HPLC), powder
SAFC
L-Arginine monohydrochloride
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
D-(+)-Glucose solution, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture