Merck
  • Tetrodotoxin abruptly blocks excitatory neurotransmission in mammalian CNS.
Tetrodotoxin abruptly blocks excitatory neurotransmission in mammalian CNS.
Toxicon : official journal of the International Society on Toxinology (2015-05-12)
Masahito Wakita, Naoki Kotani, Norio Akaike
ABSTRACT
The present study utilised a 'synaptic bouton' preparation of mechanically isolated rat hippocampal CA3 pyramidal neurons, which permits direct physiological and pharmacological quantitative analyses at the excitatory and inhibitory single synapse level. Evoked excitatory and inhibitory postsynaptic currents (eEPSCs and eIPSCs) were generated by focal paired-pulse electrical stimulation of single boutons. The sensitivity of eEPSC to tetrodotoxin (TTX) was higher than that of the voltage-dependent Na(+) channel whole-cell current (INa) in the postsynaptic CA3 soma membrane. The synaptic transmission was strongly inhibited by 3 nM TTX, at which concentration the INa was hardly suppressed. The IC50 values of eEPSC and INa for TTX were 2.8 and 37.9 nM, respectively, and complete inhibition was 3-10 nM for eEPSC and 1000 nM for INa. On the other hand, both eEPSC and eIPSC were equally and gradually inhibited by decreasing the external Na(+) concentration ([Na]o), which decreases the Na(+)gradient across the cell membrane. The results indicate that TTX at 3-10 nM could block most of voltage-dependent Na(+) channels on presynaptic nerve terminal, resulting in abruptly inhibition of action potential dependent excitatory neurotransmission.
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
CNQX, ≥98% (HPLC), solid
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
1-(2-Hydroxyethyl)piperazine, 98%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
SAFC
HEPES
SAFC
HEPES
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Adenosine, ≥99%