Merck
  • Accurate measurement of the essential micronutrients methionine, homocysteine, vitamins B6, B12, B9 and their metabolites in plasma, brain and maternal milk of mice using LC/MS ion trap analysis.
Accurate measurement of the essential micronutrients methionine, homocysteine, vitamins B6, B12, B9 and their metabolites in plasma, brain and maternal milk of mice using LC/MS ion trap analysis.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-07-22)
J Efraim Oosterink, Eva F G Naninck, Aniko Korosi, Paul J Lucassen, Johannes B van Goudoever, Henk Schierbeek
ABSTRACT
Methionine, homocysteine, vitamins B6, B12, B9, and their metabolites are crucial co-factors and substrates for many basic biological pathways including one-carbon metabolism, and they are particularly important for brain function and development and epigenetic mechanisms. These are essential nutrients that cannot be synthesized endogenously and thus need to be taken in via diet. A novel method was developed that enables simultaneous assessment of the exact concentrations of these essential micronutrients in various matrices, including maternal milk, plasma, and brain of neonatal mice. The protocol for analysis of these components in the various matrices consists of a cleanup step (i.e. lipid extraction followed by protein precipitation) combined with a liquid chromatography mass spectrometry (LC/MS) ion trap method with high sensitivity and selectivity (SRM mode). This novel method enables the measurement of these essential nutrients with good recoveries (69-117%), and high intra-day (<10%) and high intra-day precision (defined as <15% for compounds with an isotopologue and <20% for compounds without an isotopologue as internal standard) in plasma, maternal milk, and brain of mice at low and high levels. In addition, lower limits of quantitation (LOQ) were determined for the various matrices in the range for methionine (700-2000nmol/L), homocysteine (280-460-nmol/L), vitamins B6 (5-230nmol/L), B12 (7-11nmol/L), B9 (20-30nmol/L). Degradation of vitamins and oxidation of homocysteine is limited to a minimum, and only small sample volumes (30μL plasma, 20mg brain and maternal milk) are needed for simultaneous measurement. This method can help to understand how these nutrients are transferred from mother to offspring via maternal milk, as well as how these nutrients are absorbed by the offspring and eventually taken up in various tissues amongst the brain in preclinical and clinical research settings. Therefore the method can help to explore critical periods in lactating mothers and developing offspring.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, BioReagent, for molecular biology, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Vitamin B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Vitamin B12, ≥98%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
1,4-Dithioerythritol, ≥99.0% (RT), BioUltra
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
1,4-Dithioerythritol, BioXtra, ≥99.0%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
L-Methionine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Folic acid, meets USP testing specifications
Sigma-Aldrich
Folic acid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥97%
Sigma-Aldrich
Pyridoxine, ≥98%
Sigma-Aldrich
Folic acid, ≥97%
Sigma-Aldrich
L-Methionine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Heptafluorobutyric acid, 98%
Sigma-Aldrich
Heptafluorobutyric acid, ≥99.0% (GC)
Sigma-Aldrich
L-Methionine, BioUltra, ≥99.5% (NT)
SAFC
L-Methionine
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N