Merck
  • E2-2 Dependent Plasmacytoid Dendritic Cells Control Autoimmune Diabetes.
E2-2 Dependent Plasmacytoid Dendritic Cells Control Autoimmune Diabetes.
PloS one (2015-12-02)
Lisbeth Hansen, Anja Schmidt-Christensen, Shashank Gupta, Nina Fransén-Pettersson, Tine D Hannibal, Boris Reizis, Pere Santamaria, Dan Holmberg
ABSTRACT
Autoimmune diabetes is a consequence of immune-cell infiltration and destruction of pancreatic β-cells in the islets of Langerhans. We analyzed the cellular composition of the insulitic lesions in the autoimmune-prone non-obese diabetic (NOD) mouse and observed a peak in recruitment of plasmacytoid dendritic cells (pDCs) to NOD islets around 8-9 weeks of age. This peak coincides with increased spontaneous expression of type-1-IFN response genes and CpG1585 induced production of IFN-α from NOD islets. The transcription factor E2-2 is specifically required for the maturation of pDCs, and we show that knocking out E2-2 conditionally in CD11c+ cells leads to a reduced recruitment of pDCs to pancreatic islets and reduced CpG1585 induced production of IFN-α during insulitis. As a consequence, insulitis has a less aggressive expression profile of the Th1 cytokine IFN-γ and a markedly reduced diabetes incidence. Collectively, these observations demonstrate a disease-promoting role of E2-2 dependent pDCs in the pancreas during autoimmune diabetes in the NOD mouse.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Sodium bicarbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, for molecular biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium hydrogencarbonate, −40-+140 mesh, ≥95%
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium bicarbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
Sigma-Aldrich
Sodium bicarbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ACS reagent, ≥99.7%
Sigma-Aldrich
Sodium bicarbonate, ReagentPlus®, ≥99.5%, powder