Merck
Bioinspired Breathable Architecture for Water Harvesting.
Scientific reports (2015-11-19)
Rowan M von Spreckelsen, Matthew T Harris, James M Wigzell, Rebekah C Fraser, Andrea Carletto, Daniel P K Mosquin, Douglas Justice, Jas Pal S Badyal
ABSTRACT
Thuja plicata is a coniferous tree which displays remarkable water channelling properties. In this article, an easily fabricated mesh inspired by the hierarchical macro surface structure of Thuja plicata branchlets is described which emulates this efficient water collection behaviour. The key parameters are shown to be the pore size, pore angle, mesh rotation, tilt angle (branch droop) and layering (branch overlap). Envisaged societal applications include water harvesting and low cost breathable architecture for developing countries.
Sigma-Aldrich
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluorooctyl acrylate, contains inhibitor, 97%
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Decane, anhydrous, ≥99%
Sigma-Aldrich
Decane, ≥95%
Supelco
Decane, analytical standard
Sigma-Aldrich
Tetramethylsilane, electronic grade, ≥99.99% trace metals basis
Sigma-Aldrich
Tetramethylsilane, ≥99.0% (GC)
Sigma-Aldrich
Decane, ReagentPlus®, ≥99%
Sigma-Aldrich
Tetramethylsilane, ACS reagent, NMR grade, ≥99.9%
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
2-Propanol, for molecular biology, BioReagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)