Merck
  • Increased androgen receptor gene copy number is associated with TMPRSS2-ERG rearrangement in prostatic small cell carcinoma.
Increased androgen receptor gene copy number is associated with TMPRSS2-ERG rearrangement in prostatic small cell carcinoma.
Molecular carcinogenesis (2014-04-30)
Lisha Wang, Sean R Williamson, Shaobo Zhang, Jiaoti Huang, Rodolfo Montironi, Darrell D Davison, Mingsheng Wang, Jorge L Yao, Antonio Lopez-Beltran, Adeboye O Osunkoya, Gregory T MacLennan, Lee Ann Baldridge, Xiang Du, Liang Cheng
ABSTRACT
Small cell carcinoma of the prostate (PSCC) is a highly aggressive malignancy that often develops in patients previously treated with hormonal therapy for metastatic prostatic acinar adenocarcinoma. The TMPRSS2-ERG gene rearrangement is highly specific for prostate cancer and shared by PSCC; however, the role of androgen receptor (AR) gene alterations and interaction with TMPRSS2-ERG rearrangement are incompletely understood in PSCC. Sixty-one cases of PSCC were examined for AR gene copy number and TMPRSS2-ERG rearrangement by fluorescence in situ hybridization (FISH) and AR protein expression by immunohistochemistry. Of 61 cases of PSCC, 51% (31/61) demonstrated increased AR gene copy number (FISH+), 54% (33/61) were positive for TMPRSS2-ERG gene fusion, and 38% (23/61) showed AR protein expression. Of the 31 AR FISH+ cases, 23 also showed TMPRSS2-ERG gene fusion, and 16 expressed AR protein. Of the 33 cases with TMPRSS2-ERG fusion, 28 were AR FISH+ or expressed AR protein. Statistically significant correlations were observed between AR gene copy number or AR protein expression and TMPRSS2-ERG gene fusion (P = 0.001 and P = 0.03, respectively). In summary, high AR gene copy number emerges during the development of PSCC, often in association with TMPRSS2-ERG rearrangement. This potential mechanism warrants further study. Improvement will come from understanding the biology of the disease and integrating new therapies into the treatment of this rare and aggressive tumor.
Sigma-Aldrich
Citric acid, ACS reagent, ≥99.5%
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
Citric acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Citric acid, 99%
Sigma-Aldrich
Citric acid, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Citric acid, BioUltra, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Citric acid, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture