Merck
  • Reactive Nitrogen Species Scavenging Capacity of Aqueous and Ethanolic Extracts from Galinsoga parviflora and G. quadriradiata Herbs.
Reactive Nitrogen Species Scavenging Capacity of Aqueous and Ethanolic Extracts from Galinsoga parviflora and G. quadriradiata Herbs.
Natural product communications (2016-01-12)
Marta Rogowska, Sinisa Srecec, Agnieszka Bazylko
ABSTRACT
The aim of the study was to determine the scavenging capacity of aqueous and ethanolic extracts derived from the herb of two species of Galinsoga against NO and ONOO-. In both tests the aqueous extracts of both Galinsoga species were more active than the ethanolic extracts. The highest scavenging activity against NO, and also against ONOO- was shown by G. quadriradiata aqueous extract (SC50 ± SD = 1.54 ± 0.32, and 2.10 ± 1.98, respectively). The study showed that the activity of the extracts from both Galinsoga species is related not only to the presence of flavonoids but also to caffeic acid derivatives and caffeic acid. The results of this study partly explain the beneficial effects of Galinsoga herb extracts in the treatment of skin diseases.
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications