Merck
  • Soluble PD-1 aggravates progression of collagen-induced arthritis through Th1 and Th17 pathways.
Soluble PD-1 aggravates progression of collagen-induced arthritis through Th1 and Th17 pathways.
Arthritis research & therapy (2015-11-27)
Cuiping Liu, Juean Jiang, Li Gao, Xiaoting Wang, Xiaohan Hu, Min Wu, Jian Wu, Ting Xu, Qin Shi, Xueguang Zhang
ABSTRACT
The programmed cell death 1 (PD-1) protein is a critical regulator of T-cell activation and is also an important therapeutic target for autoimmune diseases. Little is known about the regulation and functional properties of the soluble PD-1 (sPD-1) variant. The aim of this study was to examine the role of sPD-1 in the regulation of human and murine rheumatoid arthritis (RA). Expression of cytokines and sPD-1 in sera, synovial fluid, and peripheral blood (PB) mononuclear cells of patients with RA were analyzed by enzyme-linked immunosorbent assay and quantitative polymerase chain reaction. PD-1 function was assessed in PB T cells after stimulation of the cells with anti-CD3 and PD-L1-Fc to crosslink PD-1. Recombinant PD-1-Fc was injected intraperitoneally into DBA/1 mice with collagen-induced arthritis (CIA) to analyze the function of sPD-1 in vivo. High concentrations of sPD-1 were found in sera and synovial fluid of patients with RA. The levels of serum sPD-1 were significantly correlated with titers of rheumatoid factor (RF) (r = 0.306, p = 0.005) and 28-joint Disease Activity Score (r = 0.545, p < 0.001). Further characterization of sPD-1 revealed that it functionally blocked the inhibitory effect of membrane-bound PD-1 on T-cell activation. Interferon γ, tumor necrosis factor α, and interleukin 17A were identified as inducers of sPD-1 in vitro. Moreover, PD-1-Fc enhanced proinflammatory cytokine expression, generation of Th1 cells and Th17 cells, and joint pathology in a CIA model. sPD-1 regulates peripheral T-cell responses in both human and murine RA. Thus, sPD-1 may represent an additional biomarker or target in immunomodulatory therapy for RA.
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
Phorbol 12-myristate 13-acetate, ≥99% (TLC), film or powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Sulfuric acid solution, puriss. p.a., ≥25% (T)
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Sulfuric acid, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis