Merck
  • Combining inkjet printing and amorphous nanonization to prepare personalized dosage forms of poorly-soluble drugs.
Combining inkjet printing and amorphous nanonization to prepare personalized dosage forms of poorly-soluble drugs.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-09-02)
Wean Sin Cheow, Tie Yi Kiew, Kunn Hadinoto
ABSTRACT
Inkjet printing of drug nanosuspension on edible porous substrates was carried out for the first time with the objective of preparing personalized dosage forms of poorly soluble drugs. Amorphous drug-polysaccharide nanoparticle complex (or drug nanoplex in short) was used as the nanosuspension ink, instead of the conventional crystalline nanodrug. The amorphous drug nanoplex exhibited low propensity to Ostwald ripening growth, high colloidal stability, and supersaturation generation capability making it ideal for printing. Nanoplexes of ciprofloxacin - a BCS Class IV compound - prepared by complexation with dextran sulfate were used as the nanosuspension ink at two different sizes (i.e. ≈265nm and 188nm). Inkjet printing was performed on cellulose substrate at 0.25% (w/v) nanosuspension concentration and 5% (w/v) polyethylene glycol. For both nanoplex sizes, the results indicated that the printed dose could be increased by increasing the number of droplets dispensed. However, exact correlations between the achievable dose and the number of droplets dispensed were not evident, which was likely caused by the spatial non-homogeneity in the nanosuspension concentration. Compared to the larger nanoplex, printed nanodrugs of the smaller nanoplex consistently exhibited higher payload with better batch-to-batch reproducibility (<6%). The maximum achievable payload was equal to ≈2.5μg/cm(2), which was multifold higher than that achieved had inkjet printing of ciprofloxacin solution been performed. Nevertheless, print substrate with higher liquid uptake capacity is needed to increase the payload nearer to the therapeutic dose. Lastly, the drug release and non-cytotoxicity of the printed nanodrug were successfully established in vitro.
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)