Merck
  • Triacylglycerols and aliphatic alcohols from fruits of three Tunisian Pistacia lentiscus populations.
Triacylglycerols and aliphatic alcohols from fruits of three Tunisian Pistacia lentiscus populations.
Journal of the science of food and agriculture (2014-09-13)
Hajer Trabelsi, Justin Renaud, Wahid Herchi, Sadok Boukhchina, Paul Mayer
ABSTRACT
The search for other sources of vegetable oils by the exploitation and the enhancement of other oil plants will be needed to meet the demands of the international market. This study aims to determine the triacylglycerol (TAG) molecular species and aliphatic alcohol compositions of unexploited fruits of three Tunisian Pistacia lentiscus (lentisc) populations from the Korbous, Tebaba and Rimel areas of Tunisia. Results show that the content of total TAG varies from 738.32 mg g(-1) of total lipid in the Tebaba population to 981.15 mg g(-1) of total lipid in the Korbous population. Furthermore, 14 species of TAG were detected in the three studied populations. In addition, 13 aliphatic compounds were identified and classified into two groups: (1) aliphatic alcohols with fewer than 20 carbon atoms (hexadecanol, heptadecanol, (Z)-octadec-9-en-1-ol, octadecanol and nonadécanol); and (2) the policosanol group (eicosenol, docosenol, docosanol tetracosanol, hexacosanol octacosanol and triacontanol). The Tebaba population showed a distinct composition compared to Korbous and Rimel where heptadecanol is the major compound. Quantitatively, the most abundant TAG species are those constituted by palmitic, oleic and/or linoleic acid. Furthermore, the significant difference observed at the oil composition is associated with a remarkable station effect.
Sigma-Aldrich
1-Eicosanol, 98%
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, anhydrous, 99.5%
Sigma-Aldrich
Chlorotrimethylsilane solution, 1.0 M in THF
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Chlorotrimethylsilane, purified by redistillation, ≥99%
Sigma-Aldrich
Diethyl ether
Sigma-Aldrich
Chlorotrimethylsilane, Wacker Chemie AG, ≥99.0% (GC)
Sigma-Aldrich
Sodium sulfate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Diethyl ether, contains 1 ppm BHT as inhibitor, anhydrous, ≥99.7%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Chlorotrimethylsilane, ≥98.0% (GC)
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis