Merck
  • High efficiency, narrow particle size distribution, sub-2 μm based macrocyclic glycopeptide chiral stationary phases in HPLC and SFC.
High efficiency, narrow particle size distribution, sub-2 μm based macrocyclic glycopeptide chiral stationary phases in HPLC and SFC.
Analytica chimica acta (2015-11-04)
Chandan L Barhate, M Farooq Wahab, Zachary S Breitbach, David S Bell, Daniel W Armstrong
ABSTRACT
State of the art chiral chromatography still employs 3-5 μm bonded or immobilized chiral selectors in 10-25 cm columns. With the availability of 1.9 μm narrow particle size distribution (NPSD) silica, it is now possible to make ever shorter, high efficiency columns practical for sub-minute chiral separations. Three macrocyclic glycopeptides (teicoplanin, teicoplanin aglycone, and vancomycin) were bonded onto 1.9 μm NPSD particles. Such packed columns had ∼80% lower backpressure as compared to polydisperse (PD) 1.7 μm silica materials when using the same mobile phase. The decreased backpressure allowed for diminution of frictional heating and allowed for the use of the 1.9 μm NPSD particle based columns at high flow rates. The 1.9 μm NPSD particle based columns showed up to 190,000 plates m(-1) for chiral molecules and 210,000 plates m(-1) for achiral probes. Representative enantiomeric separations are shown for wide classes of compounds, including different types of amino acids, β-blockers, and pharmaceutically important heterocyclic compounds such as oxazolidinones. Applications in three liquid chromatography modes, namely, reversed phase, polar organic mode and normal phase chiral separations were shown with resolution values ranging from 1.5 to 5.7. Additionally, the same columns were used with supercritical fluid chromatography (SFC) for ultrafast separations.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
1,3-Dinitrobenzene, 97% anhydrous basis
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Pyridine, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Pyridine, ≥99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Pyridine, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Pyridine, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydrosulfite, ≥82% (RT)
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Sigma-Aldrich
Acetonitrile, for preparative HPLC, ≥99.8% (GC)
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium hydrosulfite, technical grade
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Pyridine, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)