Merck
  • Internalization of Near-Infrared Fluorescently Labeled Activatable Cell-Penetrating Peptide and of Proteins into Human Fibrosarcoma Cell Line HT-1080.
Internalization of Near-Infrared Fluorescently Labeled Activatable Cell-Penetrating Peptide and of Proteins into Human Fibrosarcoma Cell Line HT-1080.
Journal of cellular biochemistry (2014-12-30)
Felista Tansi, Eric Kallweit, Christoph Kaether, Katarina Kappe, Christina Schumann, Ingrid Hilger, Siegmund Reissmann
ABSTRACT
The internalization of near-infrared fluorescently labeled cargos into living cells and tissues allows a highly sensitive detection without interference from skin, porphins or other fluorescent cell and tissue compounds. In this study, the uptake of labeled bovine serum albumin and an antibody, into fibrosarcoma (HT-1080) cells was triggered by the formation of non-covalent complexes with different cell-penetrating peptides; uptake efficiency and intracellular localization were determined. To improve selectivity of internalization into tumor cells, a fluorescent activatable cell-penetrating peptide (ACPP) was synthesized and functionally characterized. This 25-mer peptide was designed to be activatable by Matrix-Metallo-Proteases (MMPs). Its uptake selectivity was estimated using cells with different MMP activities.
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
N,N-Dimethylformamide, for molecular biology, ≥99%
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, powder
SAFC
Glycine
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Glycine, BioUltra, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Fmoc-Arg(Pbf)-OH, ≥98.0% (HPLC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
(Benzotriazol-1-yloxy)tripyrrolidinophosphonium hexafluorophosphate, 98%
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, ≥98% (NMR)
Sigma-Aldrich
Tris(2-carboxyethyl)phosphine hydrochloride, BioUltra, suitable for electrophoresis, SDS-PAGE tested
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
4-Methylmorpholine, ReagentPlus®, 99%
Sigma-Aldrich
4-Methylmorpholine, BioXtra, suitable for protein sequencing, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
4-Methylmorpholine, purified by redistillation, ≥99.5%