Merck
  • Tissue alkaline phosphatase is involved in lipid metabolism and gene expression and secretion of adipokines in adipocytes.
Tissue alkaline phosphatase is involved in lipid metabolism and gene expression and secretion of adipokines in adipocytes.
Biochimica et biophysica acta (2015-09-24)
Claudia Hernández-Mosqueira, Cristina Velez-delValle, Walid Kuri-Harcuch
ABSTRACT
Alkaline phosphatases are dimeric hydrolytic enzymes that dephosphorylate nucleotides and proteins. AP-TNAP is found primarily in skeletal tissues were it plays a major role in the mineralization of the extracellular matrix and bone formation. In this study we found through conventional and real time PCR assays that Alpl, the gene encoding for AP-TNAP is expressed in adipose tissue and in 3 T3-F442A adipocytes. We evaluated, using RNAi its role in adipocyte metabolism, and its cytoplasmic location by immunohistochemistry. Alpl is highly expressed late in adipogenesis during adipose terminal differentiation. Knocking down Alpl increased the expression of the genes encoding for glycerophosphate dehydrogenase, and for the adipokines adiponectin, and FABP4 (aP2) but decreased that of leptin, and it also increased secretion of FABP4; these 3 proteins are important in adipocyte systemic signaling and insulin sensitivity. Inhibition of alkaline phosphatase activity in adipocytes by levamisole reduced lipolysis and the expression of various lipogenic genes. We found the enzyme intracytoplasmically, forming aggregates in close surroundings of the lipid droplets during lipolysis. We suggest that AP-TNAP activity is involved in lipid and energy metabolism of fat cells, and it might regulate glucose metabolism and insulin sensitivity via adipokine synthesis and secretion. The activity of AP-TNAP might have a critical role in the energy balance of the adipocyte, probably participating in obesity and metabolic syndrome.
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, for molecular biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
SAFC
L-Histidine
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Phe-Gly-Gly
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%