Merck
  • Functional analysis of the 11q23.3 glioma susceptibility locus implicates PHLDB1 and DDX6 in glioma susceptibility.
Functional analysis of the 11q23.3 glioma susceptibility locus implicates PHLDB1 and DDX6 in glioma susceptibility.
Scientific reports (2015-11-28)
Rebekah Baskin, Nicholas T Woods, Gustavo Mendoza-Fandiño, Peter Forsyth, Kathleen M Egan, Alvaro N A Monteiro
ABSTRACT
Glioma is the most common malignant primary brain tumor and is associated with poor prognosis. Genetic factors contributing to glioma risk have recently been investigated through genome-wide association studies (GWAS), implicating seven independent glioma risk loci in six chromosomal regions. Here, we performed an in-depth functional analysis of the risk locus proximal to the PHLDB1 gene on 11q23.3. We retrieved all SNPs in linkage disequilibrium (r(2) ≥ 0.2) with the glioma-associated SNP (rs498872) and performed a comprehensive bioinformatics and experimental functional analysis for the region. After testing candidate SNPs for allele-specific activity in a luciferase-based enhancer scanning assay, we established a subset of 10 functional SNPs in the promoters of PHLDB1 and DDX6, and in a putative enhancer element. Chromatin conformation capture (3C) identified a physical interaction between the enhancer element containing a functional SNP (rs73001406) and the promoter of the DDX6 gene. Knockdown experiments in cell culture and 3D assays to evaluate the role of PHLDB1 and DDX6 suggest that both genes may contribute to the phenotype. These studies reveal the functional landscape of the 11q23.3 glioma susceptibility locus and identify a network of functional SNPs in regulatory elements and two target genes as a possible mechanism driving glioma risk association.
Sigma-Aldrich
Ammonium hydrogen difluoride, 99.999% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, for molecular biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Magnesium chloride, anhydrous, ≥98%
Sigma-Aldrich
Magnesium chloride, BioReagent, suitable for insect cell culture, ≥97.0%
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Ammonium hydrogen difluoride, reagent grade, 95%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Phldb1
Sigma-Aldrich
MISSION® esiRNA, targeting human TREH, PHLDB1
SAFC
HEPES
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O